Learn More
Buerkle™ Aluminum Profisampler™
ProfiSampler Aluminum for solvents and flammable liquids in Explosion Categories IIA, IIB and IIC. With bottle thread GL 45 (suitable for Schott glass bottles 100 ml to 1000 ml).When taking samples of flammable liquids, the device must be earthed. For this, earthing cable and, possibly in addition, anti-static set should be used, see accessories.
Brand: Buerkle™ 5305-1100
724.84 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Alert:
Attention! Make sure to pay attention to the List of Chemical Resistance!
Description
The ProfiSampler aluminum set consists of a vacuum pump made of aluminum, with feet and venting valve and bottle thread GL 45, glass bottle 500mL, five suction tubes (black PA) 220cm long and electrically conductive, hose cutter, brass hose weight, 20 Seal-it bottle seals and an earthing cable.- Stands on three foot supports for stability and safety
- Fill level control, automatic return flow and overfill protection through built-in venting valve.
- Available with suction hose in PE, PA and PTFE wherever the highest degree of purity is required
- Can be supplied with straight suction tubes for simpler sampling, including a transport case for convenient carriage and disposal of the suction tubes
- Suction volume 200mL, suction pressure 0.5 bar
- Suction height from liquid surface 5m
- Sample bottles available in different materials and sizes
Specifications
Autoclavable |