missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Potassium Sulfate Dodecahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reagent European Pharmacopoeia,≥99.5%
37.54 € - 69.62 €
Chemical Identifiers
CAS | 7784-24-9 |
---|---|
Molecular Formula | AlH24KO20S2 |
Molecular Weight (g/mol) | 474.372 |
MDL Number | MFCD00149143 |
InChI Key | GNHOJBNSNUXZQA-UHFFFAOYSA-J |
Synonym | kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate |
PubChem CID | 62667 |
ChEBI | CHEBI:86465 |
IUPAC Name | aluminum;potassium;disulfate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15693310
|
Honeywell-Fluka
31242-500G |
500 g |
37.54 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15683310
|
Honeywell-Fluka
31242-1KG |
1 kg |
69.62 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7784-24-9 | |
474.372 | |
GNHOJBNSNUXZQA-UHFFFAOYSA-J | |
62667 | |
aluminum;potassium;disulfate;dodecahydrate |
AlH24KO20S2 | |
MFCD00149143 | |
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate | |
CHEBI:86465 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+] |
Specifications
7784-24-9 | |
AlK(SO4)2 · 12H2O | |
NONH for all modes of transport | |
GNHOJBNSNUXZQA-UHFFFAOYSA-J | |
aluminum;potassium;disulfate;dodecahydrate | |
62667 | |
474.39g/mol | |
ACS/puriss. p.a. | |
Aluminum Potassium Sulfate Dodecahydrate, Reag. Ph. Eur. |
AlH24KO20S2 | |
MFCD00149143 | |
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+] | |
474.372 | |
CHEBI:86465 | |
≥99.5% | |
1.757g/mL at 25°C |
Safety and Handling
missing translation for 'einecsNumber' : 233-141-3
missing translation for 'rtecsNumber' : WS5690000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title