Learn More
Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL, Thermo Scientific Chemicals
Used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
103.52 € - 327.63 €
Chemical Identifiers
CAS | 11145-27-0 |
---|---|
Molecular Formula | AlSi |
Molecular Weight (g/mol) | 55.067 |
MDL Number | MFCD00134029 |
InChI Key | CSDREXVUYHZDNP-UHFFFAOYSA-N |
Synonym | silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic |
PubChem CID | 9793685 |
IUPAC Name | aluminum;silicon |
SMILES | [Al].[Si] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11398997
|
Thermo Scientific Alfa Aesar
038721.AD |
50 mL |
103.52 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309007
|
Thermo Scientific Alfa Aesar
038721.AE |
100 mL |
141.71 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450171
|
Thermo Scientific Alfa Aesar
038721.AP |
500 mL |
327.63 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
11145-27-0 | |
55.067 | |
CSDREXVUYHZDNP-UHFFFAOYSA-N | |
9793685 | |
[Al].[Si] |
AlSi | |
MFCD00134029 | |
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic | |
aluminum;silicon |
Specifications
11145-27-0 | |
AlSi | |
UN3264 | |
Miscible with water. | |
[Al].[Si] | |
55.067 | |
Plastic bottle |
Liquid | |
MFCD00134029 | |
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic | |
CSDREXVUYHZDNP-UHFFFAOYSA-N | |
aluminum;silicon | |
9793685 | |
Aluminum |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8
missing translation for 'einecsNumber' : 231-714-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum, plasma standard solution, Specpure Al 10,000µg/ml
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only