missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum phosphate, pure, Thermo Scientific Chemicals
43.40 € - 489.00 €
Chemical Identifiers
CAS | 7784-30-7 |
---|---|
Molecular Formula | AlO4P |
Molecular Weight (g/mol) | 121.95 |
InChI Key | ILRRQNADMUWWFW-UHFFFAOYSA-K |
Synonym | aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1 |
PubChem CID | 64655 |
IUPAC Name | aluminum;phosphate |
SMILES | [O-]P(=O)([O-])[O-].[Al+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10061581
|
Thermo Scientific Acros
344441000 |
100 g | Glass Bottle |
43.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10215181
|
Thermo Scientific Acros
344445000 |
500 g | Glass Bottle |
153.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984291
|
Thermo Scientific Acros
344440025 |
2.5 kg | Plastic bottle |
489.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-30-7 | |
121.95 | |
aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1 | |
aluminum;phosphate |
AlO4P | |
ILRRQNADMUWWFW-UHFFFAOYSA-K | |
64655 | |
[O-]P(=O)([O-])[O-].[Al+3] |
Specifications
>1500.0°C | |
White | |
AlO4P | |
15,352 | |
Solubility in water: insoluble in water. Other solubilities: insoluble in alcohol, soluble in acids, alkali | |
[O-]P(=O)([O-])[O-].[Al+3] | |
121.95 | |
121.95 | |
Aluminum phosphate |
7784-30-7 | |
Powder | |
AlPO4 | |
aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1 | |
ILRRQNADMUWWFW-UHFFFAOYSA-K | |
aluminum;phosphate | |
64655 | |
Pure |
Safety and Handling
missing translation for 'einecsNumber' : 232-056-9
RUO – Research Use Only