Learn More
Aluminum phosphate, 97%, Thermo Scientific Chemicals
55.20 € - 143.00 €
Chemical Identifiers
CAS | 7784-30-7 |
---|---|
Molecular Formula | AlO4P |
Molecular Weight (g/mol) | 121.951 |
MDL Number | MFCD00003430 |
InChI Key | ILRRQNADMUWWFW-UHFFFAOYSA-K |
Synonym | aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1 |
PubChem CID | 64655 |
IUPAC Name | aluminum;phosphate |
SMILES | [O-]P(=O)([O-])[O-].[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366128
|
Thermo Scientific Alfa Aesar
011109.30 |
250 g |
55.20 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356128
|
Thermo Scientific Alfa Aesar
011109.A1 |
1 kg |
143.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
As a flux for ceramics, preparation of dental cement, cement with calcium sulfate and sodium silicate, special glasses, as gel in pharmacy.Aluminum phosphate is used as catalysts, molecular sieves, flux for ceramics and as a gel in pharmacy. It is employed in the preparation of dental cement, emollients and fire-resistant coating. It plays an important role as an anti-fouling agent in textile industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-30-7 | |
121.951 | |
ILRRQNADMUWWFW-UHFFFAOYSA-K | |
64655 | |
[O-]P(=O)([O-])[O-].[Al+3] |
AlO4P | |
MFCD00003430 | |
aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1 | |
aluminum;phosphate |
Specifications
>1500°C | |
White | |
97% | |
MFCD00003430 | |
Insoluble in water and acetic acid. Slightly soluble in concentrated hydrochloric acid and nitric acid.Insoluble in water. | |
[O-]P(=O)([O-])[O-].[Al+3] | |
121.951 | |
121.95 | |
2.566 g/cm3 |
7784-30-7 | |
Powder | |
AlO4P | |
aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1 | |
ILRRQNADMUWWFW-UHFFFAOYSA-K | |
aluminum;phosphate | |
64655 | |
White powder | |
Aluminum phosphate |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P315-P362-P501c
H315-H320
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'einecsNumber' : 232-056-9
missing translation for 'rtecsNumber' : TB6450000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum phosphate
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only