Learn More
Aluminum oxide, NanoDur™, AL-0450, 50% in H{2}O, colloidal dispersion with dispersant, Thermo Scientific Chemicals
Colloidal dispersion with dispersant
29.70 € - 121.00 €
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
IUPAC Name | dialuminum;oxygen(2-) |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
Aluminum oxide water borne dispersion suitable for incorporation in water compatible coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
dialuminum;oxygen(2-) |
Specifications
1344-28-1 | |
Powder | |
MFCD00003424 | |
Soluble in water. Insoluble in diethyl ether. | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
Aluminum oxide |
White | |
Al2O3 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
Odorless |
Safety and Handling
missing translation for 'tsca' : Yes
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.