missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, fused, insulating powder, 99.7+%, Thermo Scientific Chemicals
Used in Dry Filling For thermal Insulation of Electrical Furnaces
89.20 € - 293.00 €
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11383128
|
Thermo Scientific Alfa Aesar
033199.A1 |
1 kg |
89.20 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393128
|
Thermo Scientific Alfa Aesar
033199.A7 |
5 kg |
293.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in dry filling for thermal insulation of electrical furnaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Form: approx. 180 micronChemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
1344-28-1 | |
>99.7% | |
MFCD00003424 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
White powder | |
Aluminum oxide |
White | |
Al2O3 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
Odorless | |
1.33 hPa (1mmHg) at 2158°C |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'rtecsNumber' : BD1200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only