missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide desiccant, regular, Thermo Scientific Chemicals

36.68 € - 227.13 €

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11315098
View Documents
Thermo Scientific Alfa Aesar
044018.22
100 g
36.68 €
100g
Please to purchase this item. Need a web account? Register with us today!
11305098
View Documents
Thermo Scientific Alfa Aesar
044018.36
500 g
98.59 €
500g
Please to purchase this item. Need a web account? Register with us today!
11395088
View Documents
Thermo Scientific Alfa Aesar
044018.A3
2 kg
227.13 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aluminum oxide is used in dessicators ceramics, coating material and electrical insulators. It is an ingredient in sunscreen, sometimes present in cosmetics such as blush, lipstick, and nail polish. Aluminum oxide is the catalyst in the Claus process in hydro desulfurization and some Ziegler-Natta polymerizations. It is widely used as an abrasive, in the automotive industries, cosmetic industries and fiber materials.It is a medium for chromatography in some sodium vapor lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Dry gases and liquids
  • Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2
  • Liquids include: benzene, hexane, oil, pentane
Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]

Specifications

1344-28-1
Al2O3
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
Aluminum oxide desiccant, Regular
Spheres
MFCD00003424
Insoluble in water and ethanol.
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-691-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.