missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, catalyst support, low surface area, Thermo Scientific Chemicals

Product Code. 11370538
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Low surface area

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Physical Form Pellets
Quantity 5 kg
IUPAC Name dialuminium(3+) trioxidandiide
Packaging Plastic bottle
Chemical Name or Material Aluminum oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'