Learn More
Aluminum oxide, activated, neutral, gamma-phase, 99.9% (metals basis), Thermo Scientific Chemicals
81.90 € - 294.00 €
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
Aluminum oxide is used as an adsorbent, desiccating agent and in the manufacture of dental cements and refractories. It is used in protein extraction as a component in the preparation of tissues and in bushings, spindles, lapping, polishing, refractory coating for investment casting shells, decorative non-skid tile and pressure blasting. It is also used in varnishes, for textile impregnation, in cosmetics and as an intermediate in pharmaceutical production. It is a source of aluminum in various reactions. It is used as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes and as a grinding or blending agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
1344-28-1 | |
Powder | |
Al2O3 | |
14,356 | |
Insoluble in water. | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
Odorless | |
(metals basis) |
White | |
99.9% (Metals basis) | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminium(3+) trioxidandiide | |
9989226 | |
99.9% | |
Tap density: 2.4 to 5.0g/cm3 | |
Aluminum oxide, Activated, neutral, gamma-phase |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'rtecsNumber' : BD1200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only