missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide, Activated, neutral, Brockmann I, Honeywell Fluka™
Activated, neutral, Brockmann I
62.73 € - 576.16 €
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
IUPAC Name | dialuminum;oxygen(2-) |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15621720
|
Honeywell-Fluka
199974-100G |
100 g | |||||||
15631720
|
Honeywell-Fluka
199974-1KG |
1 kg |
143.90 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15641720
|
Honeywell-Fluka
199974-5KG |
5 kg |
576.16 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
dialuminum;oxygen(2-) |
Specifications
1344-28-1 | |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96g/mol |
Neutral | |
Al2O3 | |
NONH for all modes of transport | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
Aluminum Oxide, Activated Brockmann I |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'rtecsNumber' : BD1200000