missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide, Activated, basic, Brockmann I, Honeywell Fluka™
Click to view available options
Quantity:
100 g
1 kg
5 kg
20 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
pH | Basic |
Quantity | 20 kg |
Linear Formula | Al2O3 |
UN Number | NONH for all modes of transport |
Formula Weight | 101.96g/mol |
Chemical Name or Material | Aluminum Oxide, Activated Brockmann I |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'