Learn More
Gravis Aluminum Open Caps
Product Code. 11763893
Aluminum
Click to view available options
Material:
Aluminum
Providing secure closure for vessels in the laboratory. Parisienne de Verrerie™ Aluminum Open Caps are durable and resilient.
  • 40mm in diameter
  • Autoclavable
Specifications
Material Aluminum
No. per Pack 100Pack
Quantity 100 Pack
Product Type Open Top Cap
Closure Material Aluminum
Closure Type Thread 40 mm
Product Content Correction
Product Title
Gravis Aluminum Open Caps >