Learn More
Cole-Parmer™ Aluminum Oil Free Weighing Dish
Designed for moisture determination balances. Cole-Parmer™ Aluminum Oil Free Weighing Dishes are ideal for storage, dispensing, general purpose testing and weighing.
Brand:  Cole-Parmer™ 01018-20
Product Code. 11877172
  • 263.00 € / Pack of 500
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Smooth-sided, flat bottom, multipurpose dish
  • Oil-free and non-coated
  • Aluminum Oil Free 
  • Disposable 
Specifications
Specifications
Single-use
Round
Universal balances
76.2 mm
1.3 g
Aluminum
Weighing Dish
6 mm
64 mm