Learn More
Aluminum nitrate nonahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
24.60 € - 474.00 €
Chemical Identifiers
CAS | 7784-27-2 |
---|---|
Molecular Formula | AlH18N3O18 |
Molecular Weight (g/mol) | 375.129 |
MDL Number | MFCD00003419 |
InChI Key | SWCIQHXIXUMHKA-UHFFFAOYSA-N |
Synonym | aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic |
PubChem CID | 24567 |
IUPAC Name | aluminum;trinitrate;nonahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333368
|
Thermo Scientific Alfa Aesar
036291.18 |
50 g |
24.60 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323368
|
Thermo Scientific Alfa Aesar
036291.30 |
250 g |
51.60 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11313368
|
Thermo Scientific Alfa Aesar
036291.A1 |
1 kg |
114.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343368
|
Thermo Scientific Alfa Aesar
036291.D9 |
5 x 1 kg |
474.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
7784-27-2 | |
375.129 | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
24567 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
AlH18N3O18 | |
MFCD00003419 | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
aluminum;trinitrate;nonahydrate |
Specifications
73.5°C (135° (decomposition)) | |
White | |
98 -102% | |
MFCD00003419 | |
Very soluble in water and alcohol. Slightly soluble in acetone. Insoluble in ethyl acetate and pyridine. | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
375.129 | |
375.13 (212.99 Anhydrous) | |
Odorless | |
White crystalline |
7784-27-2 | |
Crystalline | |
AlH18N3O18 | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate | |
24567 | |
ACS | |
Hygroscopic | |
Aluminum nitrate nonahydrate, nonahydrate |
Safety and Handling
P280-P305+P351+P338-P310
H318
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III
missing translation for 'einecsNumber' : 236-751-8
missing translation for 'rtecsNumber' : BD1050000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
- Aluminum nitrate nonahydrate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only