missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Manganese tubing, alloy 3003, 7.14mm (0.281in) OD, 6.43mm (0.253in) ID, Thermo Scientific™
32.10 € - 98.24 €
Specifications
Chemical Name or Material | Aluminum Manganese tubing, Alloy 3003 |
---|---|
Name Note | OD: 7.14mm (0.281 in.), ID: 6.43mm (0.253 in.) |
Avg. Mol. Wt. or Mol. Wt. Range | Al:Mn:Cu; 98.6:1.2:0.12 wt % |
MDL Number | MFCD00198174 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15454025
|
Thermo Scientific Alfa Aesar
045942.RM |
90 cm | N/A | ||||||
15464025
|
Thermo Scientific Alfa Aesar
045942.RN |
5 x 90 cm |
98.24 €
90cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Aluminum Manganese tubing, Alloy 3003 | |
OD: 7.14mm (0.281 in.), ID: 6.43mm (0.253 in.) | |
Al:Mn:Cu; 98.6:1.2:0.12 wt % | |
MFCD00198174 |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures