missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Aluminum Lids for Aluminum Basin
Nickel Electro™ Aluminun Lid for aluminum Basin is crevice free and one piece construction.
45.60 € - 73.30 €
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
12787346
|
Nickel Electro™
640 |
200 mm |
45.60 €
Pack of 5 |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
12707356
|
Nickel Electro™
643 |
70 mm |
50.15 €
Pack of 5 |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
12717356
|
Nickel Electro™
644 |
75 mm |
73.30 €
Pack of 5 |
Estimated Shipment: 31-10-2024
Log in to see stock. |
|||||
12797346
|
Nickel Electro™
642 |
60 mm |
49.10 €
Pack of 5 |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
12727356
|
Nickel Electro™
646 |
90 mm |
54.05 €
Pack of 5 |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
Description
Nickel Electro™ Aluminun Lid for aluminum Basin is crevice free and one piece construction.
- Aluminum
- Seamless
- One piece construction
- Crevice free
- For use with aluminum basin
Specifications
Aluminum | |
5 mm |