missing translation for 'onlineSavingsMsg'
Learn More

Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis), Thermo Scientific Chemicals

56.98 € - 104.52 €

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316787
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010093.30
250 g
56.98 €
250g
Please to purchase this item. Need a web account? Register with us today!
11306787
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010093.A1
1 kg
104.52 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
[Al]
Al
MFCD00134029
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
CHEBI:33629

Specifications

7429-90-5
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
5359268
99.9%
Granular
Aluminum granules, 8 to 12mm (0.32 to 0.47 in.)
Al
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
[Al]
26.98
CHEBI:33629
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-072-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only