missing translation for 'onlineSavingsMsg'
Learn More

Aluminum fluoride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11400740
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum fluoride is used in the smelting process to lower the melting point of electrolytes, production of aluminum silicates, refining of aluminum scarp, production of specialty refractory products, and in glass industry as filler. It is used as flux ingredient to remove magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-18-1
Molecular Formula AlF3
MDL Number MFCD00003426

Specifications

Melting Point 1291°C (sublimation)
Physical Form Powder
Quantity 250 g
Merck Index 14,339
Solubility Information Slightly soluble in water.
Formula Weight 83.98
Percent Purity 99.99%
Sensitivity Hygroscopic
Density 2.882 g/mL
Assay (metals basis)
Chemical Name or Material Aluminum fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.