missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Fluoride, Anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11323418
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For vacuum deposition

Aluminum fluoride (AlF3) is an inorganic compound used primarily in the production of aluminum. It is also used to inhibit fermentation. It is a sputtering target for preparation of low index films. In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-18-1
MDL Number MFCD00003426

Specifications

Melting Point 1291°C (sublimation)
Physical Form Sintered Lump
Quantity 25 g
Merck Index 14,339
Solubility Information Sparingly soluble in acids and alkalies. Insoluble in Acetone.
Formula Weight 83.98
Percent Purity 99.5%
Sensitivity Hygroscopic
Density 2.882 g/mL
Chemical Name or Material Aluminum fluoride, Anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.