missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride hexahydrate, 99%, Thermo Scientific Chemicals

30.45 € - 203.01 €

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11407087
SDS View Documents
Thermo Scientific Alfa Aesar
A14437.30
250 g
30.45 €
250g
Please to purchase this item. Need a web account? Register with us today!
11497077
SDS View Documents
Thermo Scientific Alfa Aesar
A14437.0B
1000 g
48.84 €
1000g
Please to purchase this item. Need a web account? Register with us today!
11417087
SDS View Documents
Thermo Scientific Alfa Aesar
A14437.0I
5000 g
203.01 €
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

Solubility
Highly soluble in water. Soluble in alcohol, ether, glycerol, propylene glycol and acetonitrile.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
Specifications

Chemical Identifiers

7784-13-6
241.42
JGDITNMASUZKPW-UHFFFAOYSA-K
AlCl3H12O6
MFCD00149134
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

100°C (decomposition)
2.5
MFCD00149134
14,337
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminium(3+) hexahydrate trichloride
241.43 (133.34 Anhydrous)
Odorless
Aluminum chloride hexahydrate
7784-13-6
AlCl3H12O6
UN3260
Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile.
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
241.42
99%
2.398
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-208-1

missing translation for 'rtecsNumber' : BD0530000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Aluminum chloride hexahydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Aluminum chloride he

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.