missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride Hexahydrate, 99%, Reagent Grade, Honeywell™

Product Code. 15642350
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color White
pH 2.5 to 3.5 at 20°C
Quantity 500 g
UN Number NONH for all modes of transport
IUPAC Name aluminium(3+) hexahydrate trichloride
Formula Weight 241.43g/mol
Percent Purity 99%
Grade ReagentPlus™
Packaging Poly Bottle
Appearance Crystals or Crystalline Chunk(s) or Chunk(s) or Solid
Vapor Pressure 1mmHg (100°C)
Density 2.4g/cm3
Chemical Name or Material Aluminum Chloride Hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Aluminium chloride-6-hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'