missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate Hexadecahydrate, Technical, Fisher Chemical

Product Code. 10233850
2kg, Plastic powder jar
Click to view available options
2kg, Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 86°C
CAS 16828-11-8
Color White
pH 2.5
Physical Form Solid
Quantity 2 kg
Molecular Formula Al2H22O20S3
MDL Number 149137
InChI Key JANHUGNIVASISO-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.[Al].[Al].OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O
IUPAC Name tris(sulfuric acid) dialuminium octahydrate
Molecular Weight (g/mol) 492.30
Formula Weight 342.1358g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium sulfate hexadecahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum Sulfate Hexadecahydrate, Technical, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.