missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Nitrate Nonahydrate, Certified AR for Analysis, Fisher Chemical™
88.95 € - 115.00 €
Chemical Identifiers
CAS | 7784-27-2 |
---|---|
Molecular Formula | AlH18N3O18 |
Molecular Weight (g/mol) | 375.13 |
MDL Number | 149132 |
InChI Key | SWCIQHXIXUMHKA-UHFFFAOYSA-N |
IUPAC Name | aluminium(3+) nonahydrate trinitrate |
SMILES | O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
Aluminum Nitrate Nonahydrate, Certified AR for Analysis
Chemical Identifiers
7784-27-2 | |
375.13 | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
AlH18N3O18 | |
149132 | |
aluminium(3+) nonahydrate trinitrate |
Specifications
73°C | |
2.5 to 3.5 | |
AlH18N3O18 | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminium(3+) nonahydrate trinitrate | |
375.13g/mol |
7784-27-2 | |
Solid | |
149132 | |
O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
375.13 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Aluminium nitrate nonahydrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Aluminium nitrate nonahydrate, Aluminumnitrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title