missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Nitrate Nonahydrate, 98+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 10578220
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.13
MDL Number 149132
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
IUPAC Name aluminium(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 73°C
pH 2.5 to 3.5
Physical Form Solid
Quantity 5 kg
Formula Weight 375.13g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.