Learn More
Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals
68.00 € - 776.00 €
Chemical Identifiers
CAS | 7784-13-6 |
---|---|
Molecular Formula | AlCl3H12O6 |
Molecular Weight (g/mol) | 241.42 |
MDL Number | MFCD00149134 |
InChI Key | JGDITNMASUZKPW-UHFFFAOYSA-K |
SMILES | O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10492021
|
Thermo Scientific Acros
217475000 |
500 g | Glass bottle |
68.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10482021
|
Thermo Scientific Acros
217470010 |
1 kg | Glass bottle |
119.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10257313
|
Thermo Scientific Acros
217470025 |
2.5 kg | Plastic bottle |
239.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914181
|
Thermo Scientific Acros
217470100 |
10 kg | Plastic drum |
776.00 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
7784-13-6 | |
241.42 | |
JGDITNMASUZKPW-UHFFFAOYSA-K |
AlCl3H12O6 | |
MFCD00149134 | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] |
Specifications
7784-13-6 | |
AlCl3H12O6 | |
MFCD00149134 | |
15,333 | |
JGDITNMASUZKPW-UHFFFAOYSA-K | |
aluminium(3+) hexahydrate trichloride | |
241.43 | |
Pure | |
2.3900g/mL |
98.5% min. (Argentometry) | |
AlCl3·6H2O | |
01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 08,13; 09,11; 17,15 | |
Solubility in water: 477g/L (20°C). Other solubilities: soluble in alcohol, ether, glycerol and propylene | |
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-] | |
241.42 | |
99% | |
2.39 | |
Aluminum chloride hexahydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
If skin irritation occurs: Get medical advice/attention.
Wash face,hands and any ex
GHS Signal Word: Warning
- Aluminium chloride hexahydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only