missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, 99.999%, (trace metal basis), for analysis, anhydrous, powder, Thermo Scientific Chemicals

156.78 € - 564.81 €

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10364112
View Documents Promotion Details
Thermo Scientific
364810100
10 g Glass Bottle
156.78 €
10g
Please to purchase this item. Need a web account? Register with us today!
10119991
View Documents Promotion Details
Thermo Scientific Acros
364810200
20 g Glass Bottle
181.91 €
20g
Please to purchase this item. Need a web account? Register with us today!
11924181
View Documents Promotion Details
Thermo Scientific Acros
364811000
100 g Glass Bottle
564.81 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis, Lewis acid
Specifications

Chemical Identifiers

7446-70-0
133.33
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum(3+) trichloride
AlCl3
MFCD00003422
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

194.0°C
White
99.999%
AlCl3
01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum(3+) trichloride
133.34
Trace Metal Basis
2.4400g/mL
7446-70-0
Powder
AlCl3
MFCD00003422
15,333
Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform
[Al+3].[Cl-].[Cl-].[Cl-]
133.33
99.999%
2.44
Aluminum chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-208-1

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only