missing translation for 'onlineSavingsMsg'
Learn More

Aluminum ammonium sulfate dodecahydrate, 99%, pure, Thermo Scientific Chemicals

Product Code. 10134500
Click to view available options
Quantity:
5 g
100 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-26-1
MDL Number MFCD00149958
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
Synonym ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
PubChem CID 62668
IUPAC Name aluminum;azanium;disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]

Specifications

Boiling Point 200.0°C
Melting Point 93.5°C
Color White
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99%
Molecular Formula AlH28NO20S2
Linear Formula AlNH4(SO4)2·12H2O
Molecular Weight (g/mol) 453.313
Formula Weight 453.33
Percent Purity 99%
Grade Pure
Loss on Drying 45% to 48% (2 g, 300°C)
Packaging Glass Bottle
Specific Gravity 1.64
Density 1.6400g/mL
Chemical Name or Material Aluminum ammonium sulfate dodecahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.