missing translation for 'onlineSavingsMsg'
Learn More

Alumina N, Activity: Super I, MP EcoChrom™, MP Biomedicals™

MP EcoChrom™ Alumina N, Activity Super I are commonly used for purification processes of all kinds, including sample preparation for pesticides, and environmental analysis, solvent purification and all kinds of chromatography, requiring the highest precision and reproducibility.

53.00 € - 2805.00 €

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11303589
View Documents
MP Biomedicals
0204580
500 g
53.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
11313589
View Documents
MP Biomedicals
0204583
1 kg
81.20 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11323589
View Documents
MP Biomedicals
0204586
5 kg
368.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
11333589
View Documents
MP Biomedicals
0204589
50 kg
2805.00 €
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • MP EcoChrom™ Alumina N, Activity Super I shows a sorbtion capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
  • Their inital activity is kept in a very narrow range and their dactivation behavior is extremely constant.
Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]

Specifications

1344-28-1
Free-flowing Powder
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
3.94 g/cm3 (at 20°C)
Pink to White
Al2O3
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Alumina B, Activity: Super I
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-691-6

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.