missing translation for 'onlineSavingsMsg'
Learn More

Alumina B, Activity: I, MP EcoChrom™, MP Biomedicals™

Product Code. 11373519
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

MP EcoChrom™ Alumina B, Activity I are good all-round adsorbents for laboratory use as well as for industrial use. They can be adjusted to lower activity by the addition of polar media, preferably water, but also higher alcohols, (e.g. glycol or glycerol can be used).

  • MP EcoChrom™ Alumina B, Activity I is an oxide of aluminum, occurring in nature as various minerals such as bauxite, corundum, etc.
  • It is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Powder
Quantity 50 kg
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Density 3.94 g/cm3 (at 20°C)
Chemical Name or Material Alumina B, Activity: I, Activity acc. To Brockmann: 24 l/mm

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.