missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 3000 Ultrasonically Welded Model 103
Click to view available options
Size:
3X-Large
4X-Large
5X-Large
Description
- Reinforced protection: The AlphaTec™ 3000 Ultrasonically Welded - Model 103 protective suit features multi-layered fabric that ensures an effective barrier against numerous inorganic chemicals and biohazards
- Enhanced features: This protective barrier is further reinforced by its double cuffs, dual-zip systems and double zip enclosure, which extends wear life
- Increased visibility: Thanks to its highly visible bright yellow color, this protective outer garment remains particularly noticeable in low-lit environments, enhancing wearer and workplace safety
- Assured antistatic properties: In addition, it complies with EN 1149-5 antistatic standards
Specifications
Specifications
Product Type | Chemical Protective Clothing |
Size | 3X-Large |
Gender | Unisex |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Disposable | Reusable |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic, Double |
Neck Style | Collared |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction