missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 23-202 Medium Weight Double-Lined PVC Gloves

Product Code. 16845190
Click to view available options
Size:
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are constructed with both an acrylic fleece, and 13-gauge cotton liner for low temperature applications. Rough sandblast finish enhances grip capabilities in wet, slippery conditions.

  • Rough sandblast finish enhances grip in wet conditions
  • Medium duty glove for low temperature applications
  • Double lined with acrylic fleece and 13-gauge cotton
  • Designed to protect hands at temperatures as low as -30°C
  • Compliant with FDA & EU food handling requirements
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 511:2006, REACH Compliant
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish Fully Textured
Material PVC
Size 9
EN388 Standard 4231C
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KLT
EN ISO 374 - 5 Yes
Glove Cuff Style Safety
Liner Material Acrylic Fleece and 100% Cotton
Series 23-202
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'