missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Tribromoquinaldine, 97%, Thermo Scientific™

Product Code. 11430728
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-53-6
Molecular Formula C10H6Br3N
Molecular Weight (g/mol) 379.88
MDL Number MFCD00006755
InChI Key UDYYQHILRSDDMP-UHFFFAOYSA-N
PubChem CID 69178
IUPAC Name 2-(tribromomethyl)quinoline
SMILES BrC(Br)(Br)C1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 128°C to 131°C
Quantity 250 g
Beilstein 147638
Synonym 2-tribromomethyl quinoline, quinoline, 2-tribromomethyl, alpha,alpha,alpha-tribromoquinaldine, 2-alpha,alpha,alpha-tribromomethyl quinoline, .alpha.,.alpha.,.alpha.-tribromoquinaldine, acmc-209mrr, 2-tribrommethyl-chinolin, 2-tribromo-methyl-quinoline, quinoline,2-tribromomethyl, udyyqhilrsddmp-uhfffaoysa
Formula Weight 379.89
Percent Purity 97%
Chemical Name or Material alpha,alpha,alpha-Tribromoquinaldine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'