missing translation for 'onlineSavingsMsg'
Learn More

Alpha-1-Antitrypsin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen Alpha-1-Antitrypsin
Concentration 0.48 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16885984
View Documents Promotion Details
Proteintech
16382-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16875984
View Documents Promotion Details
Proteintech
16382-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The serum glycoprotein a1-AT, is a typical member of the serpin family (serine protease inhibitors), and, an inhibitor of trypsin. a1- AT is synthesized in the liver thus as a single polypeptide chain of approximately 51 kDa showing a marked degree of genetic polymorphism due to the presence of over 20 different alleles. Individuals homozygous for a1-AT defiiency are predisposed to pulmonary emphysema and hepatic cirrhosis.
Specifications

Specifications

Alpha-1-Antitrypsin
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Flow Cytometry
Unconjugated
Rabbit
Mouse, Rat, Human
P01009, P07758, P17475
20700, 24648, 5265
Alpha-1-Antitrypsin Fusion Protein Ag9369
Primary
-20°C
SERPINA1
0.48 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A1A, A1AT, AAT, Alpha 1 antiproteinase, Alpha 1 antitrypsin, Alpha 1 protease inhibitor, PI, PI1, PRO2209, PRO2275, Serpin A1, SERPINA1, SPAAT, alpha1-antitrypsin
SERPINA1, Serpina1a
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications