missing translation for 'onlineSavingsMsg'
Learn More

Allyltriethoxysilane, 97%, Thermo Scientific Chemicals

43.70 € - 159.00 €

Chemical Identifiers

CAS 2550-04-1
MDL Number MFCD00009064
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326886
View Documents
Thermo Scientific Alfa Aesar
L04297.06
5 g
43.70 €
5g
Please to purchase this item. Need a web account? Register with us today!
11336886
View Documents
Thermo Scientific Alfa Aesar
L04297.14
25 g
159.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents. Incompatible with moisture and oxidizing agents.
Specifications

Chemical Identifiers

2550-04-1
MFCD00009064

Specifications

176°C to 177°C
C9H20O3Si
MFCD00009064
1772306
allyltriethoxysilane, silane, triethoxy-2-propenyl, triethoxyallylsilane, silane, allyltriethoxy, allytriethoxysilane, triethoxy prop-2-en-1-yl silane, silane, triethoxy-2-propen-1-yl, allyl triethoxysilane, acmc-2097bb
UMFJXASDGBJDEB-UHFFFAOYSA-N
triethoxy(prop-2-enyl)silane
75692
97%
Moisture sensitive
Allyltriethoxysilane
2550-04-1
CH2=CHCH2Si(OCH2CH3)3
UN1993
21°C (69°F)
Hydrolyzes in water.
CCO[Si](CC=C)(OCC)OCC
204.34
204.34
1.407
0.953
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 219-843-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.