Learn More
Allyltrichlorosilane, 95%, Thermo Scientific Chemicals
56.60 € - 168.00 €
Chemical Identifiers
CAS | 107-37-9 |
---|---|
Molecular Formula | C3H5Cl3Si |
Molecular Weight (g/mol) | 175.508 |
MDL Number | MFCD00000486 |
InChI Key | HKFSBKQQYCMCKO-UHFFFAOYSA-N |
Synonym | allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl |
PubChem CID | 7867 |
IUPAC Name | trichloro(prop-2-enyl)silane |
SMILES | C=CC[Si](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322276
|
Thermo Scientific Alfa Aesar
L03703.06 |
5 g |
56.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332276
|
Thermo Scientific Alfa Aesar
L03703.14 |
25 g |
168.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAllyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.
Solubility
Soluble in THF, toluene, benzene. Reacts with water.
Notes
Moisture Sensitive. Store at 4°C. Store away from oxidizing agents and moisture. Incompatible with water.
Chemical Identifiers
107-37-9 | |
175.508 | |
HKFSBKQQYCMCKO-UHFFFAOYSA-N | |
7867 | |
C=CC[Si](Cl)(Cl)Cl |
C3H5Cl3Si | |
MFCD00000486 | |
allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl | |
trichloro(prop-2-enyl)silane |
Specifications
117°C to 118°C | |
C3H5Cl3Si | |
MFCD00000486 | |
1743449 | |
allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl | |
HKFSBKQQYCMCKO-UHFFFAOYSA-N | |
trichloro(prop-2-enyl)silane | |
7867 | |
95% | |
Moisture sensitive | |
Allyltrichlorosilane |
107-37-9 | |
CH2=CHCH2SiCl3 | |
UN1724 | |
18°C (64°F) | |
Soluble in THF,toluene,benzene. Reacts with water. | |
C=CC[Si](Cl)(Cl)Cl | |
175.508 | |
175.52 | |
1.449 | |
1.211 |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'einecsNumber' : 203-485-9
missing translation for 'rtecsNumber' : VV1530000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only