missing translation for 'onlineSavingsMsg'
Learn More

Allyloxytrimethylsilane, 98%, Thermo Scientific Chemicals

24.32 € - 209.04 €

Chemical Identifiers

CAS 18146-00-4
Molecular Formula C6H14OSi
Molecular Weight (g/mol) 130.262
MDL Number MFCD00008643
InChI Key MNMVKGDEKPPREK-UHFFFAOYSA-N
Synonym allyloxytrimethylsilane, allyl trimethylsilyl ether, silane, trimethyl 2-propenyloxy, trimethyl 2-propenyloxy silane, o-trimethylsilylallyl alcohol, trimethylallyloxysilane, allyloxy trimethylsilane, silane, trimethyl 2-propen-1-yloxy, trimethyl prop-2-en-1-yloxy silane, acmc-1boq9
PubChem CID 87471
IUPAC Name trimethyl(prop-2-enoxy)silane
SMILES C[Si](C)(C)OCC=C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15470258
View Documents
Thermo Scientific Alfa Aesar
H53443.06
5 g
24.32 €
5g
Please to purchase this item. Need a web account? Register with us today!
15480258
View Documents
Thermo Scientific Alfa Aesar
H53443.14
25 g
68.90 €
25g
Please to purchase this item. Need a web account? Register with us today!
15490258
View Documents
Thermo Scientific Alfa Aesar
H53443.22
100 g
209.04 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Allyloxytrimethylsilane is used in the synthesis of metalated dienes via an isomerization-elimination sequence. Allyloxytrimethylsilane [3-(Trimethylsiloxy)propene] was used in the synthesis of trimethylsilyl protected (3-hydroxypropyl)-trimethoxysilane. Used in preparation of electrical materials, OLED, and in Pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyloxytrimethylsilane is used in the synthesis of metalated dienes via an isomerization-elimination sequence. Allyloxytrimethylsilane [3-(Trimethylsiloxy)propene] was used in the synthesis of trimethylsilyl protected (3-hydroxypropyl)-trimethoxysilane. Used in preparation of electrical materials, OLED, and in Pharma industry.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
Specifications

Chemical Identifiers

18146-00-4
130.262
MNMVKGDEKPPREK-UHFFFAOYSA-N
87471
C[Si](C)(C)OCC=C
C6H14OSi
MFCD00008643
allyloxytrimethylsilane, allyl trimethylsilyl ether, silane, trimethyl 2-propenyloxy, trimethyl 2-propenyloxy silane, o-trimethylsilylallyl alcohol, trimethylallyloxysilane, allyloxy trimethylsilane, silane, trimethyl 2-propen-1-yloxy, trimethyl prop-2-en-1-yloxy silane, acmc-1boq9
trimethyl(prop-2-enoxy)silane

Specifications

101°C to 102°C
C6H14OSi
MFCD00008643
1739517
allyloxytrimethylsilane, allyl trimethylsilyl ether, silane, trimethyl 2-propenyloxy, trimethyl 2-propenyloxy silane, o-trimethylsilylallyl alcohol, trimethylallyloxysilane, allyloxy trimethylsilane, silane, trimethyl 2-propen-1-yloxy, trimethyl prop-2-en-1-yloxy silane, acmc-1boq9
C[Si](C)(C)OCC=C
130.262
130.26
Allyloxytrimethylsilane
18146-00-4
CH2=CHCH2OSi(CH3)3
UN1993
1°C (34°F)
MNMVKGDEKPPREK-UHFFFAOYSA-N
trimethyl(prop-2-enoxy)silane
87471
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only