missing translation for 'onlineSavingsMsg'
Learn More

Allylchlorodimethylsilane, 94%, Thermo Scientific Chemicals

Product Code. 11415886
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1?,4,4?-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1′,4,4′-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

Solubility
Soluble in most organic solvents; reacts with alcohols, amines, and water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, strong bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4028-23-3
Molecular Formula C5H11ClSi
Molecular Weight (g/mol) 134.678
MDL Number MFCD00000506
InChI Key KMVZWUQHMJAWSY-UHFFFAOYSA-N
Synonym allyldimethylchlorosilane, allylchlorodimethylsilane, silane, chlorodimethyl-2-propenyl, allyl chloro dimethylsilane, allyl-chloro-dimethyl silane, silane, chlorodimethyl-2-propen-1-yl, admcs, allyl chlorodimethylsilane, allylchlorodimethyl silane, acmc-1afr7
PubChem CID 77646
IUPAC Name chloro-dimethyl-prop-2-enylsilane
SMILES C[Si](C)(CC=C)Cl

Specifications

Boiling Point 110°C to 112°C
Quantity 5 g
UN Number UN2985
Beilstein 1737978
Flash Point 5°C (41°F)
Solubility Information Soluble in most organic solvents; reacts with alcohols,amines,and water.
Formula Weight 134.68
Percent Purity 97%
Refractive Index 1.429
Sensitivity Moisture sensitive
Density 0.896
Chemical Name or Material Allylchlorodimethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'