missing translation for 'onlineSavingsMsg'
Learn More

Allyl glycidyl ether, 97%, Thermo Scientific Chemicals

33.77 € - 98.99 €

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11372537
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L11634.22
100 g
33.77 €
100g
Please to purchase this item. Need a web account? Register with us today!
11382537
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L11634.36
500 g
98.99 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Allyl glycidyl ether is useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl glycidyl ether is useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

Solubility
Soluble in water (140 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

106-92-3
114.144
LSWYGACWGAICNM-UHFFFAOYSA-N
7838
C=CCOCC1CO1
C6H10O2
MFCD00005143
allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
2-(prop-2-enoxymethyl)oxirane

Specifications

106-92-3
0.967
47°C (134°F)
C6H10O2
MFCD00005143
105871
Soluble in water (140g/L at 20°C).
C=CCOCC1CO1
114.144
114.15
Allyl glycidyl ether
-100°C
50°C to 51°C (15 mmHg)
Ether-like
1.435
UN2219
allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
LSWYGACWGAICNM-UHFFFAOYSA-N
2-(prop-2-enoxymethyl)oxirane
7838
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H226-H341-H351-H361-H302-H332-H317-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P333+P313-P363-P37

H226-H302-H315-H317-H318-H331-H335-H341-H351-H361f-H373

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALLYL GLYCIDYL ETHER

missing translation for 'einecsNumber' : 203-442-4

missing translation for 'rtecsNumber' : RR0875000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only