missing translation for 'onlineSavingsMsg'
Learn More

Alizarin, 94%, Thermo Scientific Chemicals

70.75 € - 946.71 €

Chemical Identifiers

CAS 72-48-0
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001201
InChI Key RGCKGOZRHPZPFP-UHFFFAOYSA-N
Synonym alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
PubChem CID 6293
ChEBI CHEBI:16866
IUPAC Name 1,2-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11497017
View Documents
Thermo Scientific Alfa Aesar
A14404.22
100 g
70.75 €
100g
Please to purchase this item. Need a web account? Register with us today!
11407027
View Documents
Thermo Scientific Alfa Aesar
A14404.36
500 g
214.07 €
500g
Please to purchase this item. Need a web account? Register with us today!
11424240
View Documents
Thermo Scientific Alfa Aesar
A14404.0E
2500 g
946.71 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Alizarin has a major application in the manufacture of the madder lake pigments which is commonly called as Rose madder and Alizarin crimson. It is involved in the studies of bone growth, osteoporosis, bone marrow and calcium deposits in the vascular system. It is also used as a stain in calcite and aragonite to identify calcium carbonate, and synovial fluid to asses for basic calcium phosphate crystals. It is used in colorimetric measurements for quantification of amine extraction by model food simulants from epoxy polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alizarin has a major application in the manufacture of the madder lake pigments which is commonly called as Rose madder and Alizarin crimson. It is involved in the studies of bone growth, osteoporosis, bone marrow and calcium deposits in the vascular system. It is also used as a stain in calcite and aragonite to identify calcium carbonate, and synovial fluid to asses for basic calcium phosphate crystals. It is used in colorimetric measurements for quantification of amine extraction by model food simulants from epoxy polymer.

Solubility
Soluble in hexane and chloroform. Slightly soluble in water.

Notes
Incompatible with alkalis and oxidizing agents.
Specifications

Chemical Identifiers

72-48-0
240.214
RGCKGOZRHPZPFP-UHFFFAOYSA-N
6293
1,2-dihydroxyanthracene-9,10-dione
C14H8O4
MFCD00001201
alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
CHEBI:16866
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O

Specifications

72-48-0
∼430°C (sublimation)
MFCD00001201
14,251
Soluble in hexane and chloroform. Slightly soluble in water.
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3)O)O
240.214
CHEBI:16866
94%
286°C to 290°C
C14H8O4
1914037
alizarin, alizarin red, mordant red 11, 1,2-dihydroxyanthraquinone, alizarin b, turkey red, 1,2-anthraquinonediol, alizarine, alizarina, alizarine red
RGCKGOZRHPZPFP-UHFFFAOYSA-N
1,2-dihydroxyanthracene-9,10-dione
6293
240.21
Alizarin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

missing translation for 'einecsNumber' : 200-782-5

missing translation for 'rtecsNumber' : CB6580000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.