missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Alexa Fluor™ 594 Biocytin, Sodium Salt (Biocytin Alexa Fluor™ 594)
Alexa Fluor™ 594 biocytin, sodium salt (biocytin Alexa Fluor™ 594)
Brand: Molecular Probes™ A12922
385.35 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The cell-impermeant, fixable, polar tracer Alexa Fluor™ 594 biocytin combines the red-fluorescent Alexa Fluor™ 594 fluorophore with biotin and an aldehyde-fixable primary amine. Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. This water-soluble tracer can be introduced into cells by whole-cell patch clamping, iontophoresis, osmotic lysis of pinocytic vesicles or comparable methods. The biotin allows for detection and subsequent amplification with enzyme-modified streptavidins including horseradish (HRP) and alkaline phosphatase (AP) that can be used with TSA and ELF™ technologies, respectively.Specifications
594 Biocytin | |
Store at room temperature and protect from light. | |
Room Temperature | |
Alexa Fluor Dyes |
250 μg | |
Biocytins | |
Alexa Fluor™ |
For Research Use Only. Not for use in diagnostic procedures.