missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Tube, One End Closed, O.D.: 4mm, I.D.: 2mm, Thermo Scientific Chemicals
80.30 € - 180.90 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11372718
|
Thermo Scientific Alfa Aesar
030062.DM |
100 mm | N/A | ||||||
11362718
|
Thermo Scientific Alfa Aesar
030062.DR |
200 mm |
88.94 €
200mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382718
|
Thermo Scientific Alfa Aesar
030062.DW |
300 mm |
92.16 €
300mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302728
|
Thermo Scientific Alfa Aesar
030062.EH |
600 mm |
113.57 €
600mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392718
|
Thermo Scientific Alfa Aesar
030062.EM |
800 mm |
144.72 €
800mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322728
|
Thermo Scientific Alfa Aesar
030062.EO |
1000 mm |
180.90 €
1000mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Al-23 Tube | |
One end closed |
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
RUO – Research Use Only