missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Tube, Both Ends Open, O.D.: 35mm, I.D.: 29mm, Thermo Scientific Chemicals
106.53 € - 1043.19 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314017
|
Thermo Scientific Alfa Aesar
031575.DM |
100 mm |
106.53 €
100mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324017
|
Thermo Scientific Alfa Aesar
031575.DR |
200 mm | N/A | ||||||
11364017
|
Thermo Scientific Alfa Aesar
031575.DW |
300 mm |
315.57 €
300mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304017
|
Thermo Scientific Alfa Aesar
031575.EH |
600 mm | N/A | ||||||
11334017
|
Thermo Scientific Alfa Aesar
031575.EM |
800 mm | N/A | ||||||
11354017
|
Thermo Scientific Alfa Aesar
031575.EO |
1000 mm |
1043.19 €
1000mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Al-23 Tube | |
Both ends open |
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title