missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23910-1-AP-150UL
This item is not returnable.
View return policy
Description
The enzyme encoded by this gene is responsible for the catalysis of the 5-beta-reduction of bile acid intermediates and steroid hormones carrying a delta(4)-3-one structure. Deficiency of this enzyme may contribute to hepatic dysfunction.Specifications
AKR1D1 | |
Polyclonal | |
Unconjugated | |
AKR1D1 | |
3o5bred, AKR1D1, CBAS2, SRD5B1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192242, 208665, 6718 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31210, P51857, Q8VCX1 | |
AKR1D1 | |
AKR1D1 Fusion Protein Ag21009 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction