missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1B10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18252-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. This member can efficiently reduce aliphatic and aromatic aldehydes, and it is less active on hexoses. It is highly expressed in adrenal gland, small intestine, and colon, and may play an important role in liver carcinogenesis.Specifications
AKR1B10 | |
Polyclonal | |
Unconjugated | |
AKR1B10 | |
AKR1B10, AKR1B11, AKR1B12, Aldose reductase like, ALDRLn, ARL 1, ARL1, ARP, hARP, HSI, SI reductase, Small intestine reductase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296972, 57016, 67861 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60218 | |
AKR1B10 | |
AKR1B10 Fusion Protein Ag13042 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction