missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | AK1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883924
|
Proteintech
14978-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873924
|
Proteintech
14978-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenylate kinase is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate group among adinine nucleotides. Three isozymes of adenylate kinase have been identified in vertebrates, adenylate isozyme 1 (AK1), 2 (AK2) and 3 (AK3). AK1 is found in the cytosol of skeletal muscle, brain and erythrocytes, whereas AK2 and AK3 are found in the mitochondria of other tissues including liver and heart. AK1 was identified because of its association with a rare genetic disorder causing nonspherocytic hemolytic anemia where a mutation in the AK1 gene was found to reduce the catalytic activity of the enzyme.Specifications
AK1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P00568, P39069, Q9R0Y5 | |
11636, 203, 24183 | |
AK1 Fusion Protein Ag6890 | |
Primary | |
-20°C | |
AK1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
adenylate kinase 1, Adenylate kinase isoenzyme 1, AK 1, AK1, ATP AMP transphosphorylase 1, Myokinase | |
AK1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |