missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGO2 Mouse anti-Human, Unconjugated, Clone: 1G2H12, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | AGO2 |
---|---|
Clone | 1G2H12 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17561446
|
Proteintech
67934-1-IG-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17551446
|
Proteintech
67934-1-IG-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The AGO2 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human AGO2. This antibody recognizes human, mouse, rat antigen. The AGO2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
AGO2 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse, Rat | |
IgG1 |
1G2H12 | |
Monoclonal | |
Mouse | |
27161 | |
Primary |