missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Electrophoresis Grade, MP Biomedicals™

Electroendosmosis: 0.13; Gel Strength: ≥600 gm/cm2; Gel Temperature: 36°C; This is a standard agarose with good gel strength and clarity, suitable for all general applications

510.00 € - 1585.00 €

Chemical Identifiers

CAS 9012-36-6
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11471682
View Documents
MP Biomedicals
04820721
100 g
510.00 €
100g
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
 
11481682
View Documents
MP Biomedicals
04820723
500 g
1585.00 €
500g
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Chemical Identifiers

9012-36-6

Specifications

9012-36-6
Electrophoresis
Agarose
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.