missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, for biochemistry, pure powder (Mr = -0.17)

79.40 € - 968.82 €

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10154860
SDS View Documents
Thermo Scientific Chemicals
239020250
25 g Glass Bottle
79.40 €
25g
Please to purchase this item. Need a web account? Register with us today!
10472825
SDS View Documents
Thermo Scientific Chemicals
239021000
100 g Glass Bottle
241.20 €
100g
Please to purchase this item. Need a web account? Register with us today!
10380304
SDS View Documents
Thermo Scientific Chemicals
239025000
500 g Glass Bottle
968.82 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Powder
White
34.5° to 37.5°C (1.5% sol.)
Authentic
Solubility in water: soluble in hot water. Other solubilities: insoluble in most common organic solvents
Agarose, Pure powder (Mr = - 0.17), For biochemistry
9012-36-6
0.16 to 0.19 (-Mr)
>2000g/cm2 (1.5% gel)
MFCD00081294
Biochemical
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Agarose, for biochemistry, pure powder (Mr = -0.17)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.