missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals AEBSF hydrochloride, 98%

55.98 € - 487.43 €

Chemical Identifiers

CAS 30827-99-7
Molecular Formula C8H10FNO2S·ClH
Molecular Weight (g/mol) 239.7
MDL Number MFCD00132962
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
PubChem CID 186136
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10317851
View Documents
Thermo Scientific Chemicals
328110500
50 mg Glass bottle
55.98 €
50mg
Please to purchase this item. Need a web account? Register with us today!
10742885
View Documents
Thermo Scientific Chemicals
328110010
1 g Glass bottle
487.43 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

30827-99-7
239.7
WRDABNWSWOHGMS-UHFFFAOYSA-N
186136
C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl
C8H10FNO2S·ClH
MFCD00132962
4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride

Specifications

30827-99-7
100.0
White
98%
MFCD00132962
Solubility in water: 10g/L (20°C). Other solubilities: soluble in absolute ethanol
C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl
239.7
239.7
Powder
97.5
175.0°C to 177.0°C
Authentic
C8H10FNO2S·ClH
4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
WRDABNWSWOHGMS-UHFFFAOYSA-N
4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
186136
98%
AEBSF hydrochloride, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

Product Identifier
  • AEBSF hydrochloride

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only